Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144941.1 | NP_001138413.1 | 1 - 2 | 281 | 50 | |
GenBank mRNA | BC038467.1 | - | 1 - 2 | 280 | 50 | |
BI836994.1 | - | 1 - 2 | 281 | 50 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |