Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034940.hg.1 |
TC02002137.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034940.hg.1 |
ENST00000393437.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034940.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034940.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200200104.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200200104.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200200104.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200200104.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200200104.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200200104.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200200104.hg.1 |
TC0200013638.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000393437.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
ENST00000409647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013638.hg.1 |
NM_198461.3 |
Refseq |