Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148962.4 | NP_683765.1 | 1 | 561 | 69 | |
GenBank mRNA | AB083055.2 | - | 1 | 517 | 69 | |
AY158687.1 | - | 1 | 362 | 69 | ||
BC063549.1 | - | 1 | 561 | 4777971 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |