Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002629.hg.1 |
TC03000142.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002629.hg.1 |
ENST00000383768.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002629.hg.1 |
ENST00000421010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002629.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000420223.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000420223.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000420223.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000420223.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000420223.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000420223.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000420223.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000428875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000428875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000428875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000428875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000428875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000428875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000457897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000457897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000457897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000457897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000457897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
ENST00000457897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002630.hg.1 |
TC03000142.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002630.hg.1 |
ENST00000383768.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002630.hg.1 |
ENST00000421010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002630.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002632.hg.1 |
TC03000142.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002632.hg.1 |
ENST00000383768.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002632.hg.1 |
ENST00000421010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002632.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148079.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148079.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148079.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148079.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148079.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148079.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148079.hg.1 |
TC0300006898.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300148074.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148074.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148074.hg.1 |
ENST00000428875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148074.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148074.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148074.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148074.hg.1 |
ENST00000428875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148074.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148074.hg.1 |
TC0300006898.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300148078.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148078.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148078.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148078.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148078.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148078.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148078.hg.1 |
TC0300006898.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000383768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000419130.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000420223.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000420223.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000420223.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000420223.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000420223.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000420223.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000420223.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000428875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000428875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000428875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000428875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000428875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000428875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000457897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000457897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000457897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000457897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000457897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
ENST00000457897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006898.hg.1 |
NM_001040432.3 |
Refseq |