Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
ENST00000622865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
ENST00000622865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
ENST00000622865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
ENST00000622865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
ENST00000622865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
ENST00000622865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046037.hg.1 |
TC01002960.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046037.hg.1 |
ENST00000351050.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046037.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213355.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213355.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213355.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213355.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213355.hg.1 |
TC0100015206.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100213356.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213356.hg.1 |
ENST00000622865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213356.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213356.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213356.hg.1 |
ENST00000622865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213356.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213356.hg.1 |
TC0100015206.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
ENST00000351050.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
ENST00000622865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
ENST00000622865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
ENST00000622865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
ENST00000622865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
ENST00000622865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
ENST00000622865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015206.hg.1 |
NM_005272.3 |
Refseq |