Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010336.hg.1 |
ENST00000270645.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010336.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010336.hg.1 |
TC19000738.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000593483.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000593483.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000593483.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000593644.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000593644.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000593644.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000597801.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000597801.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000597801.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000597801.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000597801.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000597801.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000597801.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000598833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000598833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000598833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000598833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000598833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000598833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
ENST00000598833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010335.hg.1 |
ENST00000270645.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010335.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010335.hg.1 |
TC19000738.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010333.hg.1 |
ENST00000270645.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010333.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010333.hg.1 |
TC19000738.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900166182.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166182.hg.1 |
ENST00000597801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166182.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900166182.hg.1 |
TC1900008544.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900166182.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166182.hg.1 |
ENST00000597801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166182.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900166183.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166183.hg.1 |
ENST00000593644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166183.hg.1 |
ENST00000597801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166183.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900166183.hg.1 |
TC1900008544.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900166183.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166183.hg.1 |
ENST00000593644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166183.hg.1 |
ENST00000597801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166183.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900166185.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166185.hg.1 |
ENST00000597801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166185.hg.1 |
ENST00000598833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166185.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900166185.hg.1 |
TC1900008544.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900166185.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166185.hg.1 |
ENST00000597801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166185.hg.1 |
ENST00000598833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166185.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900166184.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166184.hg.1 |
ENST00000593644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166184.hg.1 |
ENST00000597801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166184.hg.1 |
ENST00000598833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166184.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900166184.hg.1 |
TC1900008544.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900166184.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166184.hg.1 |
ENST00000593644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166184.hg.1 |
ENST00000597801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166184.hg.1 |
ENST00000598833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166184.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900166178.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166178.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900166178.hg.1 |
TC1900008544.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900166178.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900166178.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000270645.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000593483.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000593483.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000593483.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000593644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000593644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000593644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000597801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000597801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000597801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000597801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000597801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000597801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000597801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000598833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000598833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000598833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000598833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000598833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000598833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
ENST00000598833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008544.hg.1 |
NM_020650.2 |
Refseq |