Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015136.2 | NP_055951.2 | 24 - 25 | 2707 | 118 | |
XM_005264973.3 | XP_005265030.1 | 24 - 25 | 2709 | 118 | ||
XM_005264974.2 | XP_005265031.1 | 24 - 25 | 2709 | 118 | ||
XM_006713065.1 | XP_006713128.1 | 24 - 25 | 2709 | 118 | ||
XM_017005998.1 | XP_016861487.1 | 24 - 25 | 2709 | 118 | ||
XM_017005999.1 | XP_016861488.1 | 24 - 25 | 2709 | 118 | ||
XM_017006000.1 | XP_016861489.1 | 24 - 25 | 2709 | 118 | ||
XM_017006001.1 | XP_016861490.1 | 24 - 25 | 2709 | 118 | ||
XM_017006002.1 | XP_016861491.1 | 24 - 25 | 2709 | 118 | ||
XM_017006003.1 | XP_016861492.1 | 24 - 25 | 2709 | 118 | ||
XM_017006004.1 | XP_016861493.1 | 24 - 25 | 2709 | 118 | ||
GenBank mRNA | AB052956.1 | - | 24 - 25 | 2694 | 118 | |
AJ275213.1 | - | 24 - 25 | 2663 | 118 | ||
BC150250.1 | - | 24 - 25 | 2713 | 9007475 | 118 | |
D87433.2 | - | 24 - 25 | 2692 | 118 | ||
DQ786774.1 | - | 24 - 25 | 2657 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |