Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019090.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019090.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019090.hg.1 |
TC12001459.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019091.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019091.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019091.hg.1 |
TC12001459.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000548445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000548445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000548445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000548445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000548445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000550607.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000550607.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000550607.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000550607.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000550607.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000551243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000551243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000551243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000551243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000551243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000551243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000553260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000553260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000553260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000553260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
ENST00000553260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010598.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000548445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000548445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000548445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000548445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000548445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000550607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000550607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000550607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000550607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000550607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000551243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000551243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000551243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000551243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000551243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000551243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000553260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000553260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000553260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000553260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
ENST00000553260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010598.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180303.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180303.hg.1 |
ENST00000551243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180303.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180303.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180303.hg.1 |
ENST00000551243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180303.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180303.hg.1 |
TC1200010598.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200180300.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180300.hg.1 |
ENST00000551243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180300.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180300.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180300.hg.1 |
ENST00000551243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180300.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180300.hg.1 |
TC1200010598.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200180302.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180302.hg.1 |
ENST00000551243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180302.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180302.hg.1 |
ENST00000309739.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180302.hg.1 |
ENST00000551243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180302.hg.1 |
NM_014470.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180302.hg.1 |
TC1200010598.hg.1 |
Clariom D TC |