Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0800144293.hg.1 |
ENST00000421627.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800144293.hg.1 |
ENST00000520901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800144293.hg.1 |
ENST00000522499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800144293.hg.1 |
ENST00000612087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800144293.hg.1 |
NM_001277161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800144293.hg.1 |
NM_004745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800144293.hg.1 |
ENST00000421627.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800144293.hg.1 |
ENST00000520901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800144293.hg.1 |
ENST00000522499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800144293.hg.1 |
ENST00000612087.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800144293.hg.1 |
NM_001277161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800144293.hg.1 |
NM_004745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800144293.hg.1 |
TC0800006475.hg.1 |
Clariom D TC |