Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012043.hg.1 |
ENST00000421056.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012043.hg.1 |
ENST00000421056.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012043.hg.1 |
ENST00000421056.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012043.hg.1 |
ENST00000421056.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012043.hg.1 |
ENST00000595581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012043.hg.1 |
ENST00000595581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012043.hg.1 |
ENST00000595581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008876.hg.1 |
ENST00000441512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008876.hg.1 |
ENST00000441512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008876.hg.1 |
ENST00000441512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008876.hg.1 |
ENST00000441512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008876.hg.1 |
ENST00000441512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900208085.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900208085.hg.1 |
TC1900012043.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900208085.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900208088.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900208088.hg.1 |
ENST00000421056.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900208088.hg.1 |
TC1900012043.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900208088.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900208088.hg.1 |
ENST00000421056.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900208086.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900208086.hg.1 |
TC1900012043.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900208086.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900208090.hg.1 |
ENST00000421056.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900208090.hg.1 |
TC1900012043.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900208090.hg.1 |
ENST00000421056.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008876.hg.1 |
ENST00000441512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008876.hg.1 |
ENST00000441512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008876.hg.1 |
ENST00000441512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008876.hg.1 |
ENST00000441512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008876.hg.1 |
ENST00000441512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012043.hg.1 |
ENST00000291759.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012043.hg.1 |
ENST00000421056.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012043.hg.1 |
ENST00000421056.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012043.hg.1 |
ENST00000421056.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012043.hg.1 |
ENST00000421056.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012043.hg.1 |
ENST00000595581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012043.hg.1 |
ENST00000595581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012043.hg.1 |
ENST00000595581.1 |
Ensembl |