Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004675.hg.1 |
TC07000308.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004675.hg.1 |
ENST00000435803.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004675.hg.1 |
NM_152701.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700152053.hg.1 |
ENST00000435803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152053.hg.1 |
ENST00000544596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152053.hg.1 |
ENST00000611776.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152053.hg.1 |
NM_152701.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700152053.hg.1 |
ENST00000435803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152053.hg.1 |
ENST00000544596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152053.hg.1 |
ENST00000611776.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152053.hg.1 |
NM_152701.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700152053.hg.1 |
TC0700007479.hg.1 |
Clariom D TC |