Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
ENST00000542939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
ENST00000542939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
ENST00000542939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
ENST00000542939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
NM_002567.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
NM_002567.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
NM_002567.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
NM_002567.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
NM_002567.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
NM_002567.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
NM_002567.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
NM_002567.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
NM_002567.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
NM_002567.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009071.hg.1 |
NM_002567.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167069.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167069.hg.1 |
ENST00000542939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167069.hg.1 |
NM_002567.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167069.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167069.hg.1 |
ENST00000542939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167069.hg.1 |
NM_002567.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167069.hg.1 |
TC1200009071.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
ENST00000261313.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
ENST00000542939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
ENST00000542939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
ENST00000542939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
ENST00000542939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
NM_002567.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
NM_002567.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
NM_002567.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
NM_002567.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
NM_002567.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
NM_002567.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
NM_002567.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
NM_002567.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
NM_002567.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
NM_002567.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009071.hg.1 |
NM_002567.3 |
Refseq |