Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023203.hg.1 |
ENST00000301873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023203.hg.1 |
ENST00000301874.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023203.hg.1 |
ENST00000322147.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023203.hg.1 |
ENST00000536982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023203.hg.1 |
NM_001130144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023203.hg.1 |
NM_001164266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023203.hg.1 |
NM_021070.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023203.hg.1 |
TC11001934.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100186148.hg.1 |
ENST00000301873.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186148.hg.1 |
ENST00000322147.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186148.hg.1 |
ENST00000526927.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186148.hg.1 |
ENST00000528516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186148.hg.1 |
ENST00000528966.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186148.hg.1 |
ENST00000529764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186148.hg.1 |
ENST00000530866.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186148.hg.1 |
ENST00000536982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186148.hg.1 |
NM_001130144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100186148.hg.1 |
NM_001164266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100186148.hg.1 |
NM_021070.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100186148.hg.1 |
ENST00000301873.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186148.hg.1 |
ENST00000322147.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186148.hg.1 |
ENST00000526927.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186148.hg.1 |
ENST00000528516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186148.hg.1 |
ENST00000528966.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186148.hg.1 |
ENST00000529764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186148.hg.1 |
ENST00000530866.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186148.hg.1 |
ENST00000536982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100186148.hg.1 |
NM_001130144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100186148.hg.1 |
NM_001164266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100186148.hg.1 |
NM_021070.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100186148.hg.1 |
TC1100011234.hg.1 |
Clariom D TC |