Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABGAP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014857.4 | NP_055672.3 | 18 - 19 | 2481 | 81 | |
XM_005245680.1 | XP_005245737.1 | 18 - 19 | 2411 | 81 | ||
XM_005245681.2 | XP_005245738.1 | 18 - 19 | 2370 | 81 | ||
XM_006711693.1 | XP_006711756.1 | 18 - 19 | 2411 | 81 | ||
XM_011510223.2 | XP_011508525.1 | 19 - 20 | 2438 | 81 | ||
XM_017002994.1 | XP_016858483.1 | 18 - 19 | 2368 | 81 | ||
GenBank mRNA | AB019490.1 | - | 3 - 4 | 314 | 81 | |
AB019491.1 | - | 3 - 4 | 280 | 81 | ||
AB019492.2 | - | 2 - 3 | 169 | 81 | ||
AB019493.1 | - | 3 - 4 | 511 | 81 | ||
AB449904.1 | - | 17 - 18 | 2204 | 81 | ||
AK095838.2 | - | 18 - 19 | 2378 | 81 | ||
AY364260.1 | - | 11 - 12 | 1163 | 81 | ||
BC018630.2 | - | 2 - 3 | 180 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |