Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02025896.hg.1 |
TC02001615.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02025896.hg.1 |
ENST00000407540.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02025896.hg.1 |
ENST00000421259.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02025896.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000421259.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000421259.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000421259.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000421259.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000421259.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000421259.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000421259.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000421259.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000421259.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000478482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000478482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
ENST00000478482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200186349.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200186349.hg.1 |
ENST00000421259.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200186349.hg.1 |
ENST00000478482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200186349.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200186349.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200186349.hg.1 |
ENST00000421259.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200186349.hg.1 |
ENST00000478482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200186349.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200186349.hg.1 |
TC0200011888.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000407540.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000421259.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000421259.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000421259.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000421259.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000421259.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000421259.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000421259.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000421259.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000421259.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000478482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000478482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
ENST00000478482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011888.hg.1 |
NM_002381.4 |
Refseq |