Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000051.3 | NP_000042.3 | 45 - 46 | 6954 | 104 | |
XM_005271561.4 | XP_005271618.2 | 46 - 47 | 7015 | 104 | ||
XM_005271562.4 | XP_005271619.2 | 45 - 46 | 7088 | 104 | ||
XM_006718843.3 | XP_006718906.1 | 45 - 46 | 6721 | 104 | ||
XM_006718845.1 | XP_006718908.1 | 20 - 21 | 2594 | 104 | ||
XM_011542840.2 | XP_011541142.1 | 46 - 47 | 6809 | 104 | ||
XM_011542842.2 | XP_011541144.1 | 44 - 45 | 6763 | 104 | ||
XM_011542843.2 | XP_011541145.1 | 45 - 46 | 7302 | 104 | ||
XM_011542844.2 | XP_011541146.1 | 45 - 46 | 6904 | 104 | ||
XM_011542845.1 | XP_011541147.1 | 37 - 38 | 5417 | 104 | ||
XM_017017789.1 | XP_016873278.1 | 46 - 47 | 7512 | 104 | ||
XM_017017790.1 | XP_016873279.1 | 45 - 46 | 6909 | 104 | ||
GenBank mRNA | AK299843.1 | - | 7 - 8 | 785 | 104 | |
BC137169.1 | - | 19 - 20 | 2592 | 9021166 | 104 | |
CR749436.1 | - | 1973 | 104 | |||
U26455.1 | - | 19 - 20 | 2784 | 104 | ||
U33841.1 | - | 46 - 47 | 6758 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Mammalian Gene Collection Homo sapiens Probe spans exons |