Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000502921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000502921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000502921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000502921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000502921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000502921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000502921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000502921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000503958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000503958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000503958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000503958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000503958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017108.hg.1 |
TC05001205.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017108.hg.1 |
ENST00000264463.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017108.hg.1 |
ENST00000502921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017108.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000502921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000502921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000502921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000502921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000502921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000502921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000502921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000502921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000503958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000503958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000503958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000503958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000503958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010308.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500171885.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500171885.hg.1 |
ENST00000502921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500171885.hg.1 |
ENST00000503958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500171885.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500171885.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500171885.hg.1 |
ENST00000264463.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500171885.hg.1 |
ENST00000502921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500171885.hg.1 |
ENST00000503958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500171885.hg.1 |
ENST00000510477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500171885.hg.1 |
NM_006727.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500171885.hg.1 |
TC0500010308.hg.1 |
Clariom D TC |