Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006097.hg.1 |
ENST00000016913.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006097.hg.1 |
ENST00000537076.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006097.hg.1 |
NM_001164470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006097.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006097.hg.1 |
TC11000511.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000525951.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000525951.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000525951.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000525951.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000526784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000526784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000526784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000526784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000526784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000530007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000530007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000530007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000530007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000530007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000537076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000537076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000537076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000537076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000537076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000537076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000537076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
ENST00000537076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
NM_001164470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
NM_001164470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
NM_001164470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
NM_001164470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
NM_001164470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
NM_001164470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
NM_001164470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
NM_001164470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007772.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006098.hg.1 |
ENST00000016913.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006098.hg.1 |
ENST00000537076.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006098.hg.1 |
NM_001164470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006098.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006098.hg.1 |
TC11000511.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100154877.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154877.hg.1 |
ENST00000526784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154877.hg.1 |
ENST00000537076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154877.hg.1 |
NM_001164470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154877.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154877.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154877.hg.1 |
ENST00000526784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154877.hg.1 |
ENST00000537076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154877.hg.1 |
NM_001164470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154877.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154877.hg.1 |
TC1100007772.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100154878.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154878.hg.1 |
ENST00000526784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154878.hg.1 |
ENST00000537076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154878.hg.1 |
NM_001164470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154878.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154878.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154878.hg.1 |
ENST00000526784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154878.hg.1 |
ENST00000537076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154878.hg.1 |
NM_001164470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154878.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154878.hg.1 |
TC1100007772.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000016913.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000525951.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000525951.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000525951.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000525951.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000526784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000526784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000526784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000526784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000526784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000530007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000530007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000530007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000530007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000530007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000537076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000537076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000537076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000537076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000537076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000537076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000537076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
ENST00000537076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
NM_001164470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
NM_001164470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
NM_001164470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
NM_001164470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
NM_001164470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
NM_001164470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
NM_001164470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
NM_001164470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
NM_017716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007772.hg.1 |
NM_017716.2 |
Refseq |