Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056076.hg.1 |
TC01003653.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056076.hg.1 |
ENST00000367434.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056076.hg.1 |
NM_003783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056074.hg.1 |
TC01003653.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056074.hg.1 |
ENST00000367434.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056074.hg.1 |
NM_003783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056075.hg.1 |
TC01003653.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056075.hg.1 |
ENST00000367434.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056075.hg.1 |
NM_003783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016794.hg.1 |
ENST00000367434.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016794.hg.1 |
ENST00000367434.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016794.hg.1 |
ENST00000367434.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016794.hg.1 |
ENST00000367434.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016794.hg.1 |
ENST00000367434.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016794.hg.1 |
NM_003783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016794.hg.1 |
NM_003783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016794.hg.1 |
NM_003783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016794.hg.1 |
NM_003783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016794.hg.1 |
NM_003783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227170.hg.1 |
ENST00000367434.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227170.hg.1 |
NM_003783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227170.hg.1 |
ENST00000367434.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227170.hg.1 |
NM_003783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227170.hg.1 |
TC0100016794.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100227171.hg.1 |
ENST00000367434.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227171.hg.1 |
NM_003783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227171.hg.1 |
ENST00000367434.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227171.hg.1 |
NM_003783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227171.hg.1 |
TC0100016794.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100227172.hg.1 |
ENST00000367434.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227172.hg.1 |
NM_003783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227172.hg.1 |
ENST00000367434.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227172.hg.1 |
NM_003783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227172.hg.1 |
TC0100016794.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100016794.hg.1 |
ENST00000367434.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016794.hg.1 |
ENST00000367434.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016794.hg.1 |
ENST00000367434.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016794.hg.1 |
ENST00000367434.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016794.hg.1 |
ENST00000367434.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016794.hg.1 |
NM_003783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016794.hg.1 |
NM_003783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016794.hg.1 |
NM_003783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016794.hg.1 |
NM_003783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016794.hg.1 |
NM_003783.3 |
Refseq |