Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032944.hg.1 |
TC03001821.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032944.hg.1 |
ENST00000260803.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032944.hg.1 |
ENST00000505015.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032944.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000460271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000460271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000460271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000463982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000463982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000463982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000463982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000463982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000477557.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000477557.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000477557.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000477557.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000477557.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
ENST00000477557.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000460271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000460271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000460271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000463982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000463982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000463982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000463982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000463982.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000477557.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000477557.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000477557.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000477557.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000477557.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
ENST00000477557.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012567.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300193937.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300193937.hg.1 |
ENST00000477557.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300193937.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300193937.hg.1 |
ENST00000260803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300193937.hg.1 |
ENST00000477557.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300193937.hg.1 |
NM_016216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300193937.hg.1 |
TC0300012567.hg.1 |
Clariom D TC |