Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026545.hg.1 |
TC03003413.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026545.hg.1 |
ENST00000266041.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026545.hg.1 |
ENST00000346281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026545.hg.1 |
ENST00000406595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026545.hg.1 |
ENST00000434759.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026545.hg.1 |
ENST00000485816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026545.hg.1 |
ENST00000538421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026545.hg.1 |
NM_001166449.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026545.hg.1 |
NM_002218.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207974.hg.1 |
ENST00000266041.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207974.hg.1 |
ENST00000406595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207974.hg.1 |
ENST00000441637.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207974.hg.1 |
ENST00000461966.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207974.hg.1 |
ENST00000481977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207974.hg.1 |
ENST00000485816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207974.hg.1 |
ENST00000491663.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207974.hg.1 |
ENST00000537897.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207974.hg.1 |
NM_001166449.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207974.hg.1 |
NM_002218.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207974.hg.1 |
ENST00000266041.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207974.hg.1 |
ENST00000406595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207974.hg.1 |
ENST00000441637.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207974.hg.1 |
ENST00000461966.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207974.hg.1 |
ENST00000481977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207974.hg.1 |
ENST00000485816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207974.hg.1 |
ENST00000491663.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207974.hg.1 |
ENST00000537897.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207974.hg.1 |
NM_001166449.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207974.hg.1 |
NM_002218.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207974.hg.1 |
TC0300013994.hg.1 |
Clariom D TC |