Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022450.hg.1 |
TC07001499.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022450.hg.1 |
ENST00000339594.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022450.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022450.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000466844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000466844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000466844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
ENST00000466844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000466844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000466844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000466844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
ENST00000466844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011493.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181533.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181533.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181533.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181533.hg.1 |
ENST00000339594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181533.hg.1 |
ENST00000404251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181533.hg.1 |
NM_032408.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181533.hg.1 |
TC0700011493.hg.1 |
Clariom D TC |