Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016100.hg.1 |
TC0X001191.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016100.hg.1 |
ENST00000357749.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016100.hg.1 |
ENST00000537751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016100.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016099.hg.1 |
TC0X001191.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016099.hg.1 |
ENST00000357749.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016099.hg.1 |
ENST00000537751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016099.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000467744.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000467744.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000467744.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000483950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000483950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000483950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000487515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000487515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000487515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000615443.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000615443.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000615443.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000615443.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
ENST00000615443.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
NM_001145414.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
NM_001145414.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
NM_001145414.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
NM_001145414.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010211.hg.1 |
NM_001145414.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169673.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169673.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169673.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169673.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169673.hg.1 |
TC0X00010211.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000467744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000467744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000467744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000483950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000483950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000483950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000487515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000487515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000487515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000615443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000615443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000615443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000615443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
ENST00000615443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
NM_001145414.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
NM_001145414.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
NM_001145414.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
NM_001145414.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010211.hg.1 |
NM_001145414.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169674.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169674.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169674.hg.1 |
ENST00000357749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169674.hg.1 |
NM_000390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169674.hg.1 |
TC0X00010211.hg.1 |
Clariom D TC |