Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03020446.hg.1 |
TC03001192.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03020446.hg.1 |
ENST00000285018.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03020446.hg.1 |
NM_004625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010350.hg.1 |
ENST00000285018.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010350.hg.1 |
ENST00000285018.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010350.hg.1 |
ENST00000285018.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010350.hg.1 |
ENST00000285018.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010350.hg.1 |
ENST00000285018.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010350.hg.1 |
ENST00000285018.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010350.hg.1 |
ENST00000285018.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010350.hg.1 |
ENST00000285018.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010350.hg.1 |
ENST00000285018.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010350.hg.1 |
ENST00000285018.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010350.hg.1 |
ENST00000489346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010350.hg.1 |
ENST00000489346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010350.hg.1 |
ENST00000497808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010350.hg.1 |
ENST00000497808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010350.hg.1 |
ENST00000497808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010350.hg.1 |
ENST00000497808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010350.hg.1 |
NM_004625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010350.hg.1 |
NM_004625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010350.hg.1 |
NM_004625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010350.hg.1 |
NM_004625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010350.hg.1 |
NM_004625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010350.hg.1 |
NM_004625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010350.hg.1 |
NM_004625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300175431.hg.1 |
ENST00000285018.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300175431.hg.1 |
NM_004625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300175431.hg.1 |
ENST00000285018.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300175431.hg.1 |
NM_004625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300175431.hg.1 |
TC0300010350.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300010350.hg.1 |
ENST00000285018.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010350.hg.1 |
ENST00000285018.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010350.hg.1 |
ENST00000285018.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010350.hg.1 |
ENST00000285018.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010350.hg.1 |
ENST00000285018.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010350.hg.1 |
ENST00000285018.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010350.hg.1 |
ENST00000285018.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010350.hg.1 |
ENST00000285018.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010350.hg.1 |
ENST00000285018.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010350.hg.1 |
ENST00000285018.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010350.hg.1 |
ENST00000489346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010350.hg.1 |
ENST00000489346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010350.hg.1 |
ENST00000497808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010350.hg.1 |
ENST00000497808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010350.hg.1 |
ENST00000497808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010350.hg.1 |
ENST00000497808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010350.hg.1 |
NM_004625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010350.hg.1 |
NM_004625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010350.hg.1 |
NM_004625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010350.hg.1 |
NM_004625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010350.hg.1 |
NM_004625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010350.hg.1 |
NM_004625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010350.hg.1 |
NM_004625.3 |
Refseq |