Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025401.hg.1 |
ENST00000269383.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025401.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025401.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025401.hg.1 |
TC17001888.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000540128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000540128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000540128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000540128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000540128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000540128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000540128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000540128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000540812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000540812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000540812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000540812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000591668.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000591668.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000591668.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000591668.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000591668.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000591668.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000591668.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000592642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000592642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
ENST00000592642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195813.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195813.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195813.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195813.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195813.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195813.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195813.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195813.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195813.hg.1 |
TC1700011775.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700195815.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195815.hg.1 |
ENST00000540128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195815.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195815.hg.1 |
ENST00000591668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195815.hg.1 |
ENST00000592642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195815.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195815.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195815.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195815.hg.1 |
ENST00000540128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195815.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195815.hg.1 |
ENST00000591668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195815.hg.1 |
ENST00000592642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195815.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195815.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195815.hg.1 |
TC1700011775.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700195814.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195814.hg.1 |
ENST00000540128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195814.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195814.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195814.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195814.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195814.hg.1 |
ENST00000540128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195814.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195814.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195814.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195814.hg.1 |
TC1700011775.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000269383.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000540128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000540128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000540128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000540128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000540128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000540128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000540128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000540128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000540812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000540812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000540812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000540812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000543309.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000591668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000591668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000591668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000591668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000591668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000591668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000591668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000592642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000592642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
ENST00000592642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_001256124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011775.hg.1 |
NM_173547.3 |
Refseq |