Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020787.hg.1 |
TC09001638.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020787.hg.1 |
ENST00000291900.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020787.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000414921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000414921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000414921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000414921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000414921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000414921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000427848.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000427848.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000427848.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000427848.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000494461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000494461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000494461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
ENST00000494461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020786.hg.1 |
TC09001638.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020786.hg.1 |
ENST00000291900.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020786.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000414921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000414921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000414921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000414921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000414921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000414921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000427848.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000427848.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000427848.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000427848.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000494461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000494461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000494461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
ENST00000494461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011655.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900179724.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900179724.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900179724.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900179724.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900179724.hg.1 |
TC0900011655.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900179726.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900179726.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900179726.hg.1 |
ENST00000291900.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900179726.hg.1 |
NM_006336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900179726.hg.1 |
TC0900011655.hg.1 |
Clariom D TC |