Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018070.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018070.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12018070.hg.1 |
TC12001392.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178564.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178564.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178564.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178564.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178564.hg.1 |
TC1200010413.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
ENST00000308666.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010413.hg.1 |
NM_005164.3 |
Refseq |