Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCN7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002976.3 | NP_002967.2 | 19 - 20 | 3264 | 86 | |
NR_045628.1 | 20 - 21 | 3358 | 86 | |||
XM_006712680.2 | XP_006712743.1 | 19 - 20 | 3333 | 86 | ||
XM_006712681.3 | XP_006712744.1 | 20 - 21 | 3558 | 86 | ||
XM_006712682.3 | XP_006712745.1 | 21 - 22 | 3683 | 86 | ||
XM_011511615.2 | XP_011509917.1 | 21 - 22 | 3629 | 86 | ||
XM_017004667.1 | XP_016860156.1 | 21 - 22 | 3602 | 86 | ||
GenBank mRNA | BC141864.1 | - | 20 - 21 | 3360 | 86 | |
M91556.1 | - | 19 - 20 | 3273 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Homo sapiens Probe spans exons |