Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015396.hg.1 |
TC06001281.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015396.hg.1 |
ENST00000011619.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015396.hg.1 |
ENST00000283152.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015396.hg.1 |
ENST00000539980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015396.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015397.hg.1 |
TC06001281.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015397.hg.1 |
ENST00000011619.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015397.hg.1 |
ENST00000283152.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015397.hg.1 |
ENST00000539980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015397.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015400.hg.1 |
TC06001281.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015400.hg.1 |
ENST00000011619.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015400.hg.1 |
ENST00000283152.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015400.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000469916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000469916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000469916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000469916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000469916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015401.hg.1 |
TC06001281.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015401.hg.1 |
ENST00000011619.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015401.hg.1 |
ENST00000283152.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015401.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173845.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173845.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173845.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173845.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173845.hg.1 |
TC0600010864.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600173848.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173848.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173848.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173848.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173848.hg.1 |
TC0600010864.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600173849.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173849.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173849.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173849.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173849.hg.1 |
TC0600010864.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600173846.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173846.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173846.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173846.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173846.hg.1 |
TC0600010864.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000469916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000469916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000469916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000469916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000469916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173850.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173850.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173850.hg.1 |
ENST00000011619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173850.hg.1 |
NM_005493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173850.hg.1 |
TC0600010864.hg.1 |
Clariom D TC |