Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182791.3 | NP_877592.2 | 7 - 8 | 868 | 81 | |
XM_005254692.1 | XP_005254749.1 | 17 - 18 | 2471 | 81 | ||
XM_011522085.2 | XP_011520387.1 | 18 - 19 | 3316 | 81 | ||
XM_011522086.1 | XP_011520388.1 | 17 - 18 | 2164 | 81 | ||
XM_011522087.2 | XP_011520389.1 | 18 - 19 | 2302 | 81 | ||
XM_011522088.1 | XP_011520390.1 | 17 - 18 | 2172 | 81 | ||
XM_011522089.2 | XP_011520391.1 | 11 - 12 | 1462 | 81 | ||
XM_011522090.2 | XP_011520392.2 | 7 - 8 | 1166 | 81 | ||
XM_017022623.1 | XP_016878112.1 | 18 - 19 | 3317 | 81 | ||
XM_017022625.1 | XP_016878114.1 | 18 - 19 | 3316 | 81 | ||
XM_017022627.1 | XP_016878116.1 | 18 - 19 | 3316 | 81 | ||
XM_017022629.1 | XP_016878118.1 | 20 - 21 | 3057 | 81 | ||
XM_017022632.1 | XP_016878121.1 | 7 - 8 | 1001 | 81 | ||
GenBank mRNA | AK310852.1 | - | 5 - 6 | 1149 | 81 | |
BC031684.1 | - | 14 - 15 | 1681 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |