Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
ENST00000319211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
ENST00000319211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
ENST00000319211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
ENST00000319211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
ENST00000319211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
ENST00000319211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
ENST00000319211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
ENST00000319211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
ENST00000505600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
ENST00000505600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
ENST00000505600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
NM_001311313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
NM_001311313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
NM_001311313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
NM_001311313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
NM_001311313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
NM_001311313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
NM_001311313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
NM_001311313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
NM_001311313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
NM_001992.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
NM_001992.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
NM_001992.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
NM_001992.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
NM_001992.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
NM_001992.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
NM_001992.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007831.hg.1 |
NM_001992.4 |
Refseq |