Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041722.hg.1 |
TC01002659.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041722.hg.1 |
ENST00000257177.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041722.hg.1 |
ENST00000371544.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041722.hg.1 |
NM_001009881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041722.hg.1 |
NM_015269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100206261.hg.1 |
ENST00000257177.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206261.hg.1 |
ENST00000371544.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206261.hg.1 |
ENST00000497120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206261.hg.1 |
ENST00000528642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206261.hg.1 |
NM_001009881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100206261.hg.1 |
NM_015269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100206261.hg.1 |
ENST00000257177.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206261.hg.1 |
ENST00000371544.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206261.hg.1 |
ENST00000497120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206261.hg.1 |
ENST00000528642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206261.hg.1 |
NM_001009881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100206261.hg.1 |
NM_015269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100206261.hg.1 |
TC0100014203.hg.1 |
Clariom D TC |