Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006024.hg.1 |
TC01000391.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006024.hg.1 |
ENST00000373812.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006024.hg.1 |
ENST00000396232.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006024.hg.1 |
ENST00000478283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006024.hg.1 |
ENST00000541996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006024.hg.1 |
ENST00000542507.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006024.hg.1 |
NM_001172828.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006024.hg.1 |
NM_001173128.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006024.hg.1 |
NM_016258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
ENST00000373812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
ENST00000474884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
ENST00000475796.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
ENST00000476976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
ENST00000478283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
ENST00000496288.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
ENST00000541996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
ENST00000542507.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
ENST00000621244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
NM_001172828.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
NM_001173128.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
NM_016258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
ENST00000373812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
ENST00000474884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
ENST00000475796.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
ENST00000476976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
ENST00000478283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
ENST00000496288.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
ENST00000541996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
ENST00000542507.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
ENST00000621244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
NM_001172828.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
NM_001173128.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
NM_016258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100153115.hg.1 |
TC0100007570.hg.1 |
Clariom D TC |