Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039257.hg.1 |
TC02002487.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039257.hg.1 |
ENST00000194871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039257.hg.1 |
ENST00000342193.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039257.hg.1 |
ENST00000375458.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039257.hg.1 |
ENST00000392717.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039257.hg.1 |
ENST00000409184.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039257.hg.1 |
NM_014900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200207761.hg.1 |
ENST00000342193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207761.hg.1 |
ENST00000375458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207761.hg.1 |
ENST00000392717.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207761.hg.1 |
ENST00000409184.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207761.hg.1 |
ENST00000489955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207761.hg.1 |
ENST00000493868.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207761.hg.1 |
ENST00000629362.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207761.hg.1 |
NM_001278458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200207761.hg.1 |
NM_001278460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200207761.hg.1 |
NM_001278461.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200207761.hg.1 |
NM_014900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200207761.hg.1 |
ENST00000342193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207761.hg.1 |
ENST00000375458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207761.hg.1 |
ENST00000392717.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207761.hg.1 |
ENST00000409184.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207761.hg.1 |
ENST00000489955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207761.hg.1 |
ENST00000493868.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207761.hg.1 |
ENST00000629362.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207761.hg.1 |
NM_001278458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200207761.hg.1 |
NM_001278460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200207761.hg.1 |
NM_001278461.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200207761.hg.1 |
NM_014900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200207761.hg.1 |
TC0200014792.hg.1 |
Clariom D TC |