Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016831.hg.1 |
TC08001307.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016831.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016831.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
ENST00000426346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
ENST00000426346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
ENST00000426346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
ENST00000426346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010746.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172950.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172950.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172950.hg.1 |
ENST00000276594.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172950.hg.1 |
NM_024504.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172950.hg.1 |
TC0800010746.hg.1 |
Clariom D TC |