Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000461605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000461605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000461605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000461605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000461605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000461605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000487125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000487125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000487125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000487125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000487125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000487125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000487125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000487125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000487125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000487125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000487225.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000487225.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000487225.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000487225.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000487225.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000487225.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000487225.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000487225.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000487225.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000489904.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000489904.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000489904.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000489904.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
ENST00000489904.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025868.hg.1 |
TC07001675.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025868.hg.1 |
ENST00000160382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025868.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025869.hg.1 |
TC07001675.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025869.hg.1 |
ENST00000160382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025869.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025867.hg.1 |
TC07001675.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025867.hg.1 |
ENST00000160382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025867.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186307.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186307.hg.1 |
ENST00000461605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186307.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186307.hg.1 |
ENST00000487225.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186307.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186307.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186307.hg.1 |
ENST00000461605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186307.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186307.hg.1 |
ENST00000487225.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186307.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186307.hg.1 |
TC0700012011.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000461605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000461605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000461605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000461605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000461605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000461605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000487125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000487125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000487125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000487125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000487125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000487125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000487125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000487125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000487125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000487125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000487225.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000487225.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000487225.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000487225.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000487225.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000487225.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000487225.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000487225.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000487225.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000489904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000489904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000489904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000489904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
ENST00000489904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012011.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186306.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186306.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186306.hg.1 |
ENST00000487225.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186306.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186306.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186306.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186306.hg.1 |
ENST00000487225.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186306.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186306.hg.1 |
TC0700012011.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700186309.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186309.hg.1 |
ENST00000461605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186309.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186309.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186309.hg.1 |
ENST00000160382.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186309.hg.1 |
ENST00000461605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186309.hg.1 |
ENST00000485601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186309.hg.1 |
NM_016188.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186309.hg.1 |
TC0700012011.hg.1 |
Clariom D TC |