Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000554791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000554791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000554791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000554791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000554791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000554791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000554791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000554791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000554791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000556172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000556172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000556172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000556172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000556172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557429.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557429.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557429.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557429.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557429.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557429.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557429.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557512.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557512.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557512.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557512.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557512.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557512.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557512.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557512.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557512.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557755.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557755.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557755.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557755.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557755.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557755.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
ENST00000557755.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003372.hg.1 |
ENST00000216420.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003372.hg.1 |
ENST00000557512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003372.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003372.hg.1 |
TC14000318.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000554791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000554791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000554791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000554791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000554791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000554791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000554791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000554791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000554791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000556172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000556172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000556172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000556172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000556172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557429.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557429.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557429.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557429.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557429.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557429.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557429.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557512.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557512.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557512.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557512.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557512.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557512.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557512.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557512.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557512.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
ENST00000557755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007204.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400149948.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149948.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149948.hg.1 |
ENST00000557184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149948.hg.1 |
ENST00000557317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149948.hg.1 |
ENST00000557512.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149948.hg.1 |
ENST00000557755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149948.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400149948.hg.1 |
ENST00000216420.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149948.hg.1 |
ENST00000556216.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149948.hg.1 |
ENST00000557184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149948.hg.1 |
ENST00000557317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149948.hg.1 |
ENST00000557512.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149948.hg.1 |
ENST00000557755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149948.hg.1 |
NM_006568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400149948.hg.1 |
TC1400007204.hg.1 |
Clariom D TC |