Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000310450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000310450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000310450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000310450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000310450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000310450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000310450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000310450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000310450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000463154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000463154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000463154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000463154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000463154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000463154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000463154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000463154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000463154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000480550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000480550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000480550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000480550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000480550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000480550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000480550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000480550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000481837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000481837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000481837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000481837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_016100.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_016100.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_016100.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_016100.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_016100.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_016100.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_016100.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_016100.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_016100.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_016100.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_181527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_181527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_181527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_181527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_181527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_181527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_181527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_181527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_181528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_181528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_181528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_181528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_181528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_181528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_181528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_181528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006867.hg.1 |
NM_181528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001727.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001727.hg.1 |
NM_181527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001727.hg.1 |
TC20000135.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001734.hg.1 |
ENST00000310450.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001734.hg.1 |
ENST00000334982.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001734.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001734.hg.1 |
ENST00000484480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001734.hg.1 |
NM_016100.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001734.hg.1 |
NM_181527.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001734.hg.1 |
NM_181528.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001734.hg.1 |
TC20000135.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000310450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000310450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000310450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000310450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000310450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000310450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000310450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000310450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000310450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000463154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000463154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000463154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000463154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000463154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000463154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000463154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000463154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000463154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000480550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000480550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000480550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000480550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000480550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000480550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000480550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000480550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000481837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000481837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000481837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000481837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_016100.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_016100.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_016100.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_016100.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_016100.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_016100.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_016100.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_016100.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_016100.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_016100.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_181527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_181527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_181527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_181527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_181527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_181527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_181527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_181527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_181528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_181528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_181528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_181528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_181528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_181528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_181528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_181528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006867.hg.1 |
NM_181528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146985.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146985.hg.1 |
ENST00000481837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146985.hg.1 |
NM_181527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146985.hg.1 |
TC2000006867.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000146985.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146985.hg.1 |
ENST00000481837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146985.hg.1 |
NM_181527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146995.hg.1 |
ENST00000310450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146995.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146995.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146995.hg.1 |
ENST00000463154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146995.hg.1 |
ENST00000480550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146995.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146995.hg.1 |
NM_016100.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146995.hg.1 |
NM_181527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146995.hg.1 |
NM_181528.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146995.hg.1 |
TC2000006867.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000146995.hg.1 |
ENST00000310450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146995.hg.1 |
ENST00000334982.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146995.hg.1 |
ENST00000398602.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146995.hg.1 |
ENST00000463154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146995.hg.1 |
ENST00000480550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146995.hg.1 |
ENST00000484480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146995.hg.1 |
NM_016100.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146995.hg.1 |
NM_181527.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146995.hg.1 |
NM_181528.3 |
Refseq |