Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015191.hg.1 |
ENST00000298295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015191.hg.1 |
ENST00000432283.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015191.hg.1 |
ENST00000496638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015191.hg.1 |
NM_007021.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015191.hg.1 |
TC10001216.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015195.hg.1 |
NM_007021.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015195.hg.1 |
TC10001216.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010482.hg.1 |
ENST00000298295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010482.hg.1 |
ENST00000298295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010482.hg.1 |
ENST00000298295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010482.hg.1 |
ENST00000298295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010482.hg.1 |
ENST00000298295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010482.hg.1 |
ENST00000298295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010482.hg.1 |
ENST00000298295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010482.hg.1 |
ENST00000448778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010482.hg.1 |
ENST00000448778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010482.hg.1 |
ENST00000448778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010482.hg.1 |
ENST00000448778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010482.hg.1 |
ENST00000448778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010482.hg.1 |
ENST00000496638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010482.hg.1 |
ENST00000496638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010482.hg.1 |
ENST00000496638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010482.hg.1 |
NM_007021.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010482.hg.1 |
NM_007021.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010482.hg.1 |
NM_007021.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010482.hg.1 |
NM_007021.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010482.hg.1 |
NM_007021.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010482.hg.1 |
NM_007021.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010482.hg.1 |
NM_007021.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015192.hg.1 |
ENST00000298295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015192.hg.1 |
ENST00000432283.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015192.hg.1 |
ENST00000496638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015192.hg.1 |
NM_007021.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015192.hg.1 |
TC10001216.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000171197.hg.1 |
ENST00000298295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171197.hg.1 |
ENST00000448778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171197.hg.1 |
ENST00000496638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171197.hg.1 |
NM_007021.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171197.hg.1 |
ENST00000298295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171197.hg.1 |
ENST00000448778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171197.hg.1 |
ENST00000496638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171197.hg.1 |
NM_007021.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171197.hg.1 |
TC1000010482.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000171198.hg.1 |
ENST00000298295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171198.hg.1 |
ENST00000448778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171198.hg.1 |
ENST00000496638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171198.hg.1 |
NM_007021.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171198.hg.1 |
ENST00000298295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171198.hg.1 |
ENST00000448778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171198.hg.1 |
ENST00000496638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171198.hg.1 |
NM_007021.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171198.hg.1 |
TC1000010482.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000010482.hg.1 |
ENST00000298295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010482.hg.1 |
ENST00000298295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010482.hg.1 |
ENST00000298295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010482.hg.1 |
ENST00000298295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010482.hg.1 |
ENST00000298295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010482.hg.1 |
ENST00000298295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010482.hg.1 |
ENST00000298295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010482.hg.1 |
ENST00000448778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010482.hg.1 |
ENST00000448778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010482.hg.1 |
ENST00000448778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010482.hg.1 |
ENST00000448778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010482.hg.1 |
ENST00000448778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010482.hg.1 |
ENST00000496638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010482.hg.1 |
ENST00000496638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010482.hg.1 |
ENST00000496638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010482.hg.1 |
NM_007021.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010482.hg.1 |
NM_007021.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010482.hg.1 |
NM_007021.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010482.hg.1 |
NM_007021.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010482.hg.1 |
NM_007021.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010482.hg.1 |
NM_007021.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010482.hg.1 |
NM_007021.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171201.hg.1 |
NM_007021.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171201.hg.1 |
NM_007021.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171201.hg.1 |
TC1000010482.hg.1 |
Clariom D TC |