Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
ENST00000482591.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
ENST00000482591.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
ENST00000482591.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
ENST00000482591.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
ENST00000482591.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
ENST00000493994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
ENST00000493994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
ENST00000493994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
ENST00000493994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011330.hg.1 |
TC01000708.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011330.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011330.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011329.hg.1 |
TC01000708.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011329.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011329.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160751.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160751.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160751.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160751.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160751.hg.1 |
TC0100008559.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100160752.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160752.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160752.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160752.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160752.hg.1 |
TC0100008559.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100160753.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160753.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160753.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160753.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160753.hg.1 |
TC0100008559.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100160750.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160750.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160750.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160750.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160750.hg.1 |
TC0100008559.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
ENST00000371129.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
ENST00000482591.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
ENST00000482591.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
ENST00000482591.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
ENST00000482591.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
ENST00000482591.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
ENST00000493994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
ENST00000493994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
ENST00000493994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
ENST00000493994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008559.hg.1 |
NM_014495.3 |
Refseq |