Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000462067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000462067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000462067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000462067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000462067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000477040.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000477040.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000477040.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000488606.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000488606.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000488606.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000488606.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000488606.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038980.hg.1 |
TC01002511.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038980.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038980.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038981.hg.1 |
TC01002511.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038981.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01038981.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202300.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202300.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202300.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202300.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202300.hg.1 |
TC0100013723.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000462067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000462067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000462067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000462067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000462067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000477040.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000477040.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000477040.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000488606.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000488606.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000488606.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000488606.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000488606.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202301.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202301.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202301.hg.1 |
ENST00000373116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202301.hg.1 |
NM_031280.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202301.hg.1 |
TC0100013723.hg.1 |
Clariom D TC |