Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
ENST00000357330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
ENST00000357330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
ENST00000357330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
ENST00000357330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
ENST00000357330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
ENST00000357330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
ENST00000357330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
ENST00000357330.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
ENST00000440823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
ENST00000440823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
ENST00000440823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
ENST00000440823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
ENST00000440823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
ENST00000440823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
ENST00000440823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
ENST00000590002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
ENST00000590002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
NM_001145546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
NM_001145546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
NM_001145546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
NM_001145546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
NM_001145546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
NM_001145546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
NM_001145546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
NM_033106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
NM_033106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
NM_033106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
NM_033106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
NM_033106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
NM_033106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
NM_033106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008946.hg.1 |
NM_033106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012648.hg.1 |
ENST00000357330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012648.hg.1 |
ENST00000440823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012648.hg.1 |
NM_001145546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012648.hg.1 |
NM_033106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012648.hg.1 |
TC19000919.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
ENST00000357330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
ENST00000357330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
ENST00000357330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
ENST00000357330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
ENST00000357330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
ENST00000357330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
ENST00000357330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
ENST00000357330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
ENST00000440823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
ENST00000440823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
ENST00000440823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
ENST00000440823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
ENST00000440823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
ENST00000440823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
ENST00000440823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
ENST00000590002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
ENST00000590002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
NM_001145546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
NM_001145546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
NM_001145546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
NM_001145546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
NM_001145546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
NM_001145546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
NM_001145546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
NM_033106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
NM_033106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
NM_033106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
NM_033106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
NM_033106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
NM_033106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
NM_033106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008946.hg.1 |
NM_033106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900170240.hg.1 |
ENST00000357330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170240.hg.1 |
ENST00000440823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170240.hg.1 |
ENST00000590002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170240.hg.1 |
NM_001145546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900170240.hg.1 |
NM_033106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900170240.hg.1 |
TC1900008946.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900170240.hg.1 |
ENST00000357330.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170240.hg.1 |
ENST00000440823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170240.hg.1 |
ENST00000590002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170240.hg.1 |
NM_001145546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900170240.hg.1 |
NM_033106.3 |
Refseq |