Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
ENST00000288861.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
ENST00000288861.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
ENST00000288861.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
ENST00000288861.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
ENST00000288861.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
ENST00000288861.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
ENST00000288861.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
ENST00000288861.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
ENST00000288861.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
ENST00000288861.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
ENST00000403670.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
ENST00000403670.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
ENST00000403670.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
ENST00000403670.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
ENST00000403670.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
ENST00000403670.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
ENST00000403670.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
ENST00000405346.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
ENST00000405346.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
ENST00000405346.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
ENST00000405346.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
ENST00000405346.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
NM_001029881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
NM_001029881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
NM_001029881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
NM_001029881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
NM_001029881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
NM_001029881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
NM_001029881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
NM_001029881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
NM_001029881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012040.hg.1 |
NM_001029881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
ENST00000288861.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
ENST00000288861.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
ENST00000288861.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
ENST00000288861.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
ENST00000288861.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
ENST00000288861.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
ENST00000288861.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
ENST00000288861.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
ENST00000288861.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
ENST00000288861.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
ENST00000403670.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
ENST00000403670.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
ENST00000403670.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
ENST00000403670.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
ENST00000403670.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
ENST00000403670.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
ENST00000403670.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
ENST00000405346.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
ENST00000405346.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
ENST00000405346.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
ENST00000405346.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
ENST00000405346.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
NM_001029881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
NM_001029881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
NM_001029881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
NM_001029881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
NM_001029881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
NM_001029881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
NM_001029881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
NM_001029881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
NM_001029881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012040.hg.1 |
NM_001029881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200187696.hg.1 |
ENST00000288861.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200187696.hg.1 |
NM_001029881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200187696.hg.1 |
ENST00000288861.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200187696.hg.1 |
NM_001029881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200187696.hg.1 |
TC0200012040.hg.1 |
Clariom D TC |