Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284301.2 | NP_001271230.1 | 8 - 9 | 1747 | 139 | |
XM_006715747.3 | XP_006715810.1 | 8 - 9 | 1744 | 139 | ||
XM_017012356.1 | XP_016867845.1 | 8 - 9 | 1744 | 139 | ||
GenBank mRNA | AK001472.1 | - | 4 - 5 | 525 | 139 | |
BC070066.1 | - | 8 - 9 | 1729 | 4825802 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |