Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036545.hg.1 |
TC02002276.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036545.hg.1 |
ENST00000259238.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036545.hg.1 |
ENST00000316724.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036545.hg.1 |
ENST00000346226.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036545.hg.1 |
ENST00000351659.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036545.hg.1 |
ENST00000352848.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036545.hg.1 |
ENST00000357970.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036545.hg.1 |
ENST00000393040.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036545.hg.1 |
ENST00000393041.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036545.hg.1 |
ENST00000409400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036545.hg.1 |
ENST00000466111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036545.hg.1 |
NM_004305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036545.hg.1 |
NM_139343.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036545.hg.1 |
NM_139344.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036545.hg.1 |
NM_139345.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036545.hg.1 |
NM_139346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036545.hg.1 |
NM_139347.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036545.hg.1 |
NM_139348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036545.hg.1 |
NM_139349.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036545.hg.1 |
NM_139350.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000259238.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000316724.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000346226.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000351659.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000352848.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000357970.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000393040.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000393041.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000409400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000462958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000466111.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000484253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
NM_004305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
NM_139343.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
NM_139344.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
NM_139345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
NM_139346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
NM_139347.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
NM_139348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
NM_139349.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
NM_139350.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000259238.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000316724.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000346226.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000351659.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000352848.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000357970.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000393040.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000393041.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000409400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000462958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000466111.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
ENST00000484253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
NM_004305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
NM_139343.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
NM_139344.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
NM_139345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
NM_139346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
NM_139347.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
NM_139348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
NM_139349.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
NM_139350.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203389.hg.1 |
TC0200014160.hg.1 |
Clariom D TC |