Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018219.hg.1 |
ENST00000286627.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018219.hg.1 |
ENST00000354353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018219.hg.1 |
ENST00000372437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018219.hg.1 |
ENST00000372440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018219.hg.1 |
ENST00000372443.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018219.hg.1 |
ENST00000404857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018219.hg.1 |
ENST00000406533.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018219.hg.1 |
ENST00000412708.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018219.hg.1 |
NM_001014797.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018219.hg.1 |
NM_001161352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018219.hg.1 |
NM_001161353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018219.hg.1 |
NM_002247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018219.hg.1 |
TC10001439.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000286627.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000286628.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000354353.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000372403.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000372408.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000372421.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000372437.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000372440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000372443.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000404771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000404857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000406533.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000434208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000457953.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000604624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000626620.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
NM_001014797.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
NM_001161352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
NM_001161353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
NM_001271518.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
NM_001271519.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
NM_002247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000286627.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000286628.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000354353.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000372403.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000372408.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000372421.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000372437.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000372440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000372443.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000404771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000404857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000406533.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000434208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000457953.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000604624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
ENST00000626620.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
NM_001014797.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
NM_001161352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
NM_001161353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
NM_001271518.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
NM_001271519.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
NM_002247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175740.hg.1 |
TC1000011114.hg.1 |
Clariom D TC |