Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008867.hg.1 |
TC07000597.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008867.hg.1 |
ENST00000326775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008867.hg.1 |
ENST00000451158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008867.hg.1 |
ENST00000537357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008867.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008872.hg.1 |
TC07000597.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008872.hg.1 |
ENST00000326775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008872.hg.1 |
ENST00000394170.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008872.hg.1 |
ENST00000439985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008872.hg.1 |
ENST00000451158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008872.hg.1 |
ENST00000537357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008872.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008872.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008498.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700159439.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159439.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159439.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159439.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159439.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700159439.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700159439.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159439.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159439.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159439.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159439.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700159439.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700159439.hg.1 |
TC0700008498.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000394170.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
ENST00000454175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
NM_014569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008498.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700159434.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159434.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159434.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700159434.hg.1 |
ENST00000326775.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159434.hg.1 |
ENST00000451158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159434.hg.1 |
NM_145102.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700159434.hg.1 |
TC0700008498.hg.1 |
Clariom D TC |