Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02000329.hg.1 |
TC02000024.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02000329.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02000329.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000471711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000471711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000471711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000471711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000471711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000471711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000476389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000476389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000476389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000476389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000476389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000476389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000476389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000476389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000476389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
ENST00000476389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200144844.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200144844.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200144844.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200144844.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200144844.hg.1 |
TC0200006539.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000252505.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000471711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000471711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000471711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000471711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000471711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000471711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000476389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000476389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000476389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000476389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000476389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000476389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000476389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000476389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000476389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
ENST00000476389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006539.hg.1 |
NM_018436.3 |
Refseq |