Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002888.hg.1 |
ENST00000324229.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002888.hg.1 |
ENST00000523376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002888.hg.1 |
ENST00000533448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002888.hg.1 |
NM_000728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002888.hg.1 |
TC11000215.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002887.hg.1 |
ENST00000324229.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002887.hg.1 |
ENST00000533448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002887.hg.1 |
NM_000728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002887.hg.1 |
TC11000215.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012969.hg.1 |
ENST00000324229.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012969.hg.1 |
ENST00000324229.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012969.hg.1 |
ENST00000324229.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012969.hg.1 |
ENST00000324229.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012969.hg.1 |
ENST00000324229.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012969.hg.1 |
ENST00000324229.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012969.hg.1 |
ENST00000324229.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012969.hg.1 |
ENST00000324229.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012969.hg.1 |
ENST00000533448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012969.hg.1 |
ENST00000533448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012969.hg.1 |
ENST00000533448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012969.hg.1 |
ENST00000533448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012969.hg.1 |
ENST00000533448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012969.hg.1 |
ENST00000533448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012969.hg.1 |
ENST00000533448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012969.hg.1 |
NM_000728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012969.hg.1 |
NM_000728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012969.hg.1 |
NM_000728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012969.hg.1 |
NM_000728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012969.hg.1 |
NM_000728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012969.hg.1 |
NM_000728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012969.hg.1 |
NM_000728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100199842.hg.1 |
ENST00000324229.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100199842.hg.1 |
ENST00000533448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100199842.hg.1 |
NM_000728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100199842.hg.1 |
ENST00000324229.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100199842.hg.1 |
ENST00000533448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100199842.hg.1 |
NM_000728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100199842.hg.1 |
TC1100012969.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100012969.hg.1 |
ENST00000324229.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012969.hg.1 |
ENST00000324229.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012969.hg.1 |
ENST00000324229.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012969.hg.1 |
ENST00000324229.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012969.hg.1 |
ENST00000324229.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012969.hg.1 |
ENST00000324229.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012969.hg.1 |
ENST00000324229.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012969.hg.1 |
ENST00000324229.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012969.hg.1 |
ENST00000533448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012969.hg.1 |
ENST00000533448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012969.hg.1 |
ENST00000533448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012969.hg.1 |
ENST00000533448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012969.hg.1 |
ENST00000533448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012969.hg.1 |
ENST00000533448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012969.hg.1 |
ENST00000533448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012969.hg.1 |
NM_000728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012969.hg.1 |
NM_000728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012969.hg.1 |
NM_000728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012969.hg.1 |
NM_000728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012969.hg.1 |
NM_000728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012969.hg.1 |
NM_000728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012969.hg.1 |
NM_000728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100199840.hg.1 |
ENST00000324229.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100199840.hg.1 |
ENST00000533448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100199840.hg.1 |
NM_000728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100199840.hg.1 |
ENST00000324229.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100199840.hg.1 |
ENST00000533448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100199840.hg.1 |
NM_000728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100199840.hg.1 |
TC1100012969.hg.1 |
Clariom D TC |