Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011516998.1 | XP_011515300.1 | 1 - 2 | 87 | 61 | |
XM_011516999.1 | XP_011515301.1 | 1 - 2 | 87 | 61 | ||
GenBank mRNA | BC028197.1 | - | 1 - 2 | 167 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |